Product Details

SNP ID
rs9394145
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:33432001 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCCCGCTGCCAGCCTCTCCGCCGT[C/T]GCTGCTCTTCCTGCTGCTTTCCGGG
Phenotype
MIM: 603384
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR5004 PubMed Links

Gene Details

Gene
MIR5004
Gene Name
microRNA 5004
There are no transcripts associated with this gene.

Gene
SYNGAP1
Gene Name
synaptic Ras GTPase activating protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006772.2 Intron NP_006763.2

View Full Product Details