Product Details

SNP ID
rs9861850
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:161085454 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTTACAGCCTAGTGAGCTTATAA[C/T]TCAGTAACACCCTTTTAAGAAATTC
Phenotype
MIM: 603094 MIM: 611931
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
B3GALNT1 PubMed Links

Gene Details

Gene
B3GALNT1
Gene Name
beta-1,3-N-acetylgalactosaminyltransferase 1 (globoside blood group)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001038628.1 1929 UTR 3 NP_001033717.1
NM_003781.3 1929 UTR 3 NP_003772.1
NM_033167.2 1929 UTR 3 NP_149357.1
NM_033168.2 1929 UTR 3 NP_149358.1
NM_033169.2 1929 UTR 3 NP_149359.1
XM_005247859.4 1929 UTR 3 XP_005247916.1
XM_005247861.3 1929 UTR 3 XP_005247918.1
XM_005247862.3 1929 UTR 3 XP_005247919.1
XM_005247864.3 1929 UTR 3 XP_005247921.1
XM_006713803.3 1929 UTR 3 XP_006713866.1
XM_006713804.3 1929 UTR 3 XP_006713867.1
XM_006713805.3 1929 UTR 3 XP_006713868.1
XM_017007402.1 1929 UTR 3 XP_016862891.1
XM_017007403.1 1929 UTR 3 XP_016862892.1
XM_017007404.1 1929 UTR 3 XP_016862893.1
XM_017007405.1 1929 UTR 3 XP_016862894.1
XM_017007406.1 1929 UTR 3 XP_016862895.1
XM_017007407.1 1929 UTR 3 XP_016862896.1
XM_017007408.1 1929 UTR 3 XP_016862897.1
XM_017007409.1 1929 UTR 3 XP_016862898.1
XM_017007410.1 1929 UTR 3 XP_016862899.1
XM_017007411.1 1929 UTR 3 XP_016862900.1
XM_017007412.1 1929 UTR 3 XP_016862901.1
XM_017007413.1 1929 Intron XP_016862902.1
Gene
PPM1L
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent 1L
There are no transcripts associated with this gene.

View Full Product Details