Product Details

SNP ID
rs9508843
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:19683869 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTCCTCTTAAGATTATATGGTAA[C/T]GTTTCCTGTAAGTTATCAAAGAAAT
Phenotype
MIM: 612408
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PSPC1 PubMed Links

Gene Details

Gene
PSPC1
Gene Name
paraspeckle component 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042414.2 2420 Intron NP_001035879.1
XM_006719844.3 2420 Intron XP_006719907.1
XM_011535137.1 2420 Intron XP_011533439.1
XM_011535138.2 2420 UTR 3 XP_011533440.1
XM_011535139.1 2420 Intron XP_011533441.1
XM_011535140.1 2420 Intron XP_011533442.1
XM_011535141.2 2420 Intron XP_011533443.1
XM_011535142.2 2420 Intron XP_011533444.1
XM_017020650.1 2420 Intron XP_016876139.1

View Full Product Details