Product Details

SNP ID
rs9666593
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:68054640 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCCTAATCTAAAAGTCTGCAATT[C/T]TAGAAGCCAGGAAGATTACAATTAA
Phenotype
MIM: 118491 MIM: 604592
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CHKA PubMed Links

Gene Details

Gene
CHKA
Gene Name
choline kinase alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277.2 Intron NP_001268.2
NM_212469.1 Intron NP_997634.1
XM_017017147.1 Intron XP_016872636.1
XM_017017148.1 Intron XP_016872637.1
Gene
MIR6753
Gene Name
microRNA 6753
There are no transcripts associated with this gene.

Gene
TCIRG1
Gene Name
T-cell immune regulator 1, ATPase H+ transporting V0 subunit a3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006019.3 Intron NP_006010.2
NM_006053.3 Intron NP_006044.1
XM_005273709.3 Intron XP_005273766.1
XM_011544726.2 Intron XP_011543028.1
XM_017017089.1 Intron XP_016872578.1
XM_017017090.1 Intron XP_016872579.1

View Full Product Details