Product Details

SNP ID
rs6681293
Assay Type
Functionally tested
NCBI dbSNP Submissions
6
Location
Chr.1:177176142 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCAACAGCCAGTCCCATTATTCCTC[A/T]TACTGAGGCTCACATGAAATGCAAA
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
BRINP2 PubMed Links

Gene Details

Gene
BRINP2
Gene Name
BMP/retinoic acid inducible neural specific 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021165.3 Intron NP_066988.1
XM_005245379.1 Intron XP_005245436.1
XM_011509826.1 Intron XP_011508128.1

View Full Product Details