Product Details
- SNP ID
-
rs10027520
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:176185323 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCAGACTCTGTAGGTCACTCAGTT[C/T]TAATGGGAGGGGCCAGTGGCAACTA
- Phenotype
-
MIM: 609879
MIM: 609005
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
SPATA4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs142508946] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SPATA4
- Gene Name
- spermatogenesis associated 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_144644.3 |
|
Intron |
|
|
NP_653245.2 |
- Gene
- WDR17
- Gene Name
- WD repeat domain 17
There are no transcripts associated with this gene.
View Full Product Details