Product Details

SNP ID
rs7519755
Assay Type
Functionally tested
NCBI dbSNP Submissions
44
Location
Chr.1:56495379 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGACACTTACGGATAGAGGATGAGG[A/G]AAACTCTCTACCGAGATTTAACCCA
Phenotype
MIM: 607125
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PLPP3 PubMed Links

Gene Details

Gene
PLPP3
Gene Name
phospholipid phosphatase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003713.4 2676 UTR 3 NP_003704.3

View Full Product Details