Product Details

SNP ID
rs5982097
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:73213947 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTCCTCATTACCATACATCTCTTC[A/G]TGACACATTTCCTCATCATCACAGT
Phenotype
MIM: 300026
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NAP1L2 PubMed Links

Gene Details

Gene
NAP1L2
Gene Name
nucleosome assembly protein 1 like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021963.3 928 Silent Mutation CAC,CAT H182H NP_068798.1

View Full Product Details