Product Details

SNP ID
rs10050945
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:157629269 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTATGACAAGTACCCCATAAATAC[A/G]TAAAATGTTATATATCAATAAAATA
Phenotype
MIM: 606698
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SOX30 PubMed Links

Gene Details

Gene
SOX30
Gene Name
SRY-box 30
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308165.1 Intron NP_001295094.1
NM_007017.2 Intron NP_008948.1
NM_178424.1 Intron NP_848511.1
XM_005265803.4 Intron XP_005265860.1
XM_011534420.1 Intron XP_011532722.1

View Full Product Details