Product Details

SNP ID
rs6633808
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:136158501 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGATTTTCCTCCTCCATGTCTTTCC[C/T]GTATCCTGATGCCACTTCTCTTGCA
Phenotype
MIM: 300163
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FHL1 PubMed Links

Gene Details

Gene
FHL1
Gene Name
four and a half LIM domains 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001159699.1 Intron NP_001153171.1
NM_001159700.1 Intron NP_001153172.1
NM_001159701.1 Intron NP_001153173.1
NM_001159702.2 Intron NP_001153174.1
NM_001159703.1 Intron NP_001153175.1
NM_001159704.1 Intron NP_001153176.1
NM_001167819.1 Intron NP_001161291.1
NM_001449.4 Intron NP_001440.2
XM_006724743.2 Intron XP_006724806.1
XM_006724744.2 Intron XP_006724807.1
XM_006724745.3 Intron XP_006724808.1
XM_006724746.2 Intron XP_006724809.1
XM_006724747.2 Intron XP_006724810.1
XM_011531316.1 Intron XP_011529618.1
XM_017029357.1 Intron XP_016884846.1

View Full Product Details