Product Details

SNP ID
rs10193373
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:281886 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGATTTAAACAGAAGAAAGTCTGTG[C/T]GACACATAGGCAGTGGATGAGCTGC
Phenotype
MIM: 171500
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACP1 PubMed Links

Gene Details

Gene
ACP1
Gene Name
acid phosphatase 1, soluble
There are no transcripts associated with this gene.

Gene
FAM150B
Gene Name
family with sequence similarity 150 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002919.2 Intron NP_001002919.2
XM_005264689.4 Intron XP_005264746.1
XM_006711879.3 Intron XP_006711942.1
XM_011510342.2 Intron XP_011508644.1
Gene
LOC101927262
Gene Name
melanoma-associated antigen C1-like
There are no transcripts associated with this gene.

View Full Product Details