Product Details

SNP ID
rs6693197
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:158288753 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGCATTCAGTGAATAATTAATATGA[A/T]TTAAGAAACACTATAATAATAAATG
Phenotype
MIM: 188360 MIM: 188340
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
CD1B PubMed Links

Gene Details

Gene
CD1B
Gene Name
CD1b molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001764.2 Intron NP_001755.1
XM_011510119.2 Intron XP_011508421.1
XM_017002784.1 Intron XP_016858273.1
XM_017002785.1 Intron XP_016858274.1
XM_017002786.1 Intron XP_016858275.1
Gene
CD1C
Gene Name
CD1c molecule
There are no transcripts associated with this gene.

View Full Product Details