Product Details
- SNP ID
-
rs8182278
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:15031942 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CTTCAGTTAAATGAGCCAATAAAGT[C/G]CCTGTTTTAAAACCAGTTTGAATTG
- Phenotype
-
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
CDRT7
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs80340954] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CDRT7
- Gene Name
- CMT1A duplicated region transcript 7 (non-protein coding)
There are no transcripts associated with this gene.
View Full Product Details