Product Details
- SNP ID
-
rs7520461
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
40
- Location
-
Chr.1:31263335 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TATCACAGAATCTTTTTGTTTTTCT[A/C]AATTAATTTTTTTTTTTTTTTTTTG
- Phenotype
-
MIM: 607797
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SNRNP40
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs35912395] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SNRNP40
- Gene Name
- small nuclear ribonucleoprotein U5 subunit 40
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004814.2 |
|
Intron |
|
|
NP_004805.2 |
View Full Product Details