Product Details

SNP ID
rs9919708
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:93384648 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGGCTGCATTCAAAACCATCTTGG[A/G]CTGCATGCAGTCCACGGGCCACAGG
Phenotype
MIM: 609229
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC643339 PubMed Links

Gene Details

Gene
LOC643339
Gene Name
uncharacterized LOC643339
There are no transcripts associated with this gene.

Gene
NUDT4
Gene Name
nudix hydrolase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301022.1 Intron NP_001287951.1
NM_001301023.1 Intron NP_001287952.1
NM_001301024.1 Intron NP_001287953.1
NM_019094.5 Intron NP_061967.3
NM_199040.3 Intron NP_950241.1
XM_011537794.2 Intron XP_011536096.1
XM_011537795.2 Intron XP_011536097.1
XM_011537796.2 Intron XP_011536098.1

View Full Product Details