Product Details

SNP ID
rs10482729
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:218349220 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TATTCTGTATGGAAGTTTAGGAGCA[C/G]AAAGCCATATTAATAGCAGGTTCGA
Phenotype
MIM: 190220
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TGFB2 PubMed Links

Gene Details

Gene
TGFB2
Gene Name
transforming growth factor beta 2
There are no transcripts associated with this gene.

Gene
TGFB2-AS1
Gene Name
TGFB2 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details