Product Details

SNP ID
rs6020717
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:50735943 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGAATAAAGAAAATGGAGGAGAAA[C/T]TGTGACTTTTGATAACATCATGAGA
Phenotype
MIM: 608975
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PARD6B PubMed Links

Gene Details

Gene
PARD6B
Gene Name
par-6 family cell polarity regulator beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032521.2 Intron NP_115910.1

View Full Product Details