Product Details

SNP ID
rs8193035
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:24249151 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGTTTATTAGCATCTGTTCCCCCA[T/G]TGCAAGTGAAGACTGACTCCCTCTC
Phenotype
MIM: 610781 MIM: 190195 MIM: 604319
Polymorphism
T/G, Transversion substitution
Allele Nomenclature
Literature Links
GMPR2 PubMed Links

Gene Details

Gene
GMPR2
Gene Name
guanosine monophosphate reductase 2
There are no transcripts associated with this gene.

Gene
TGM1
Gene Name
transglutaminase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000359.2 2740 UTR 3 NP_000350.1
Gene
TINF2
Gene Name
TERF1 interacting nuclear factor 2
There are no transcripts associated with this gene.

View Full Product Details