Product Details

SNP ID
rs6641102
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:12797869 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGACATGCAATTTTCAAAAACTTA[A/G]CTGCTATTTACTCTTTCTCCAGAAG
Phenotype
MIM: 311860
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PRPS2 PubMed Links

Gene Details

Gene
PRPS2
Gene Name
phosphoribosyl pyrophosphate synthetase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039091.2 Intron NP_001034180.1
NM_002765.4 Intron NP_002756.1

View Full Product Details