Product Details

SNP ID
rs9987152
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:138133392 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTGCTTCAGGCCATGGAGGTCTTGG[A/G]GCCATGCAGTGCAATTGAATTGCTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM135B PubMed Links

Gene Details

Gene
FAM135B
Gene Name
family with sequence similarity 135 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015912.3 Intron NP_056996.2
XM_011517056.2 Intron XP_011515358.1
XM_011517058.2 Intron XP_011515360.1
XM_011517059.1 Intron XP_011515361.1
XM_011517060.2 Intron XP_011515362.1
XM_011517061.2 Intron XP_011515363.1
XM_011517062.1 Intron XP_011515364.1
XM_011517063.2 Intron XP_011515365.1
XM_011517064.2 Intron XP_011515366.1
XM_011517065.1 Intron XP_011515367.1
XM_011517068.2 Intron XP_011515370.1
XM_011517069.2 Intron XP_011515371.1
XM_011517070.2 Intron XP_011515372.1
XM_011517072.2 Intron XP_011515374.1
XM_011517073.2 Intron XP_011515375.1
XM_011517074.1 Intron XP_011515376.1
XM_017013470.1 Intron XP_016868959.1
XM_017013471.1 Intron XP_016868960.1

View Full Product Details