Product Details
- SNP ID
-
rs9322855
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:20685140 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TTTGAAGCAGGCCTTAGTTTTTTGC[A/C]AGCGTAACTAACATTTGGGGATCGG
- Phenotype
-
MIM: 105850
MIM: 601030
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ANG
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs146973333] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ANG
- Gene Name
- angiogenin
- Gene
- RNASE4
- Gene Name
- ribonuclease A family member 4
View Full Product Details