Product Details

SNP ID
rs10425682
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:42088922 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGTCTCTGCTGCCTCCGTCGCCCA[A/G]CCCAACAGTTTCATTTCCCGGGTGG
Phenotype
MIM: 164176
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
POU2F2 PubMed Links
Additional Information
For this assay, SNP(s) [rs139504468] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
POU2F2
Gene Name
POU class 2 homeobox 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001207025.2 4539 UTR 3 NP_001193954.1
NM_001207026.1 4539 Intron NP_001193955.1
NM_001247994.1 4539 UTR 3 NP_001234923.1
NM_002698.4 4539 UTR 3 NP_002689.1
XM_005259010.3 4539 UTR 3 XP_005259067.1
XM_011527040.2 4539 UTR 3 XP_011525342.1
XM_011527041.2 4539 UTR 3 XP_011525343.1
XM_011527042.2 4539 UTR 3 XP_011525344.1
XM_011527043.2 4539 UTR 3 XP_011525345.2
XM_017026884.1 4539 UTR 3 XP_016882373.1
XM_017026885.1 4539 UTR 3 XP_016882374.1
XM_017026886.1 4539 UTR 3 XP_016882375.1
XM_017026887.1 4539 UTR 3 XP_016882376.1
XM_017026888.1 4539 UTR 3 XP_016882377.1
XM_017026889.1 4539 UTR 3 XP_016882378.1
XM_017026890.1 4539 UTR 3 XP_016882379.1
XM_017026891.1 4539 UTR 3 XP_016882380.1
XM_017026892.1 4539 UTR 3 XP_016882381.1
XM_017026893.1 4539 UTR 3 XP_016882382.1
XM_017026894.1 4539 UTR 3 XP_016882383.1
XM_017026895.1 4539 UTR 3 XP_016882384.1
XM_017026896.1 4539 UTR 3 XP_016882385.1
Gene
ZNF574
Gene Name
zinc finger protein 574
There are no transcripts associated with this gene.

View Full Product Details