Product Details

SNP ID
rs10405139
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57602672 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAAGGGTTATGAAGGATGAGTGAA[C/T]ATAAGATGGATGTGGAGGCTGCATT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZIK1 PubMed Links

Gene Details

Gene
ZIK1
Gene Name
zinc finger protein interacting with K protein 1
There are no transcripts associated with this gene.

Gene
ZNF530
Gene Name
zinc finger protein 530
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321981.1 Intron NP_001308910.1
NM_020880.4 Intron NP_065931.3
XM_006723194.1 Intron XP_006723257.1
XM_011526919.2 Intron XP_011525221.1
XM_011526921.2 Intron XP_011525223.1

View Full Product Details