Product Details
- SNP ID
-
hCV30569652
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:109406386 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAATGTAACAGAATTGATGCAACAC[A/G]GGTGAATCTCAAAATCATGCCAAGT
- Phenotype
-
MIM: 176844
MIM: 602458
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PSMA5
PubMed Links
Gene Details
- Gene
- PSMA5
- Gene Name
- proteasome subunit alpha 5
There are no transcripts associated with this gene.
- Gene
- SORT1
- Gene Name
- sortilin 1
There are no transcripts associated with this gene.
View Full Product Details