Product Details

SNP ID
rs7519513
Assay Type
Functionally tested
NCBI dbSNP Submissions
17
Location
Chr.1:160610009 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAACAGCACCATCAAACATTTGCTA[C/T]GTTTTTCTTTAAAACTGAATGCCAT
Phenotype
MIM: 603492
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLAMF1 PubMed Links

Gene Details

Gene
SLAMF1
Gene Name
signaling lymphocytic activation molecule family member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003037.3 2417 UTR 3 NP_003028.1
XM_005245456.3 2417 UTR 3 XP_005245513.1
XM_011509905.2 2417 UTR 3 XP_011508207.1
XM_017002129.1 2417 UTR 3 XP_016857618.1
XM_017002130.1 2417 UTR 3 XP_016857619.1
XM_017002131.1 2417 UTR 3 XP_016857620.1

View Full Product Details