Product Details
- SNP ID
-
rs6901377
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:6346529 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- CAATGACATCTCAAGCAAGATGTGA[C/T]AACAGTTACACAAAGCAGTCTACAG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
LY86-AS1
PubMed Links
Gene Details
- Gene
- LY86-AS1
- Gene Name
- LY86 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details