Product Details

SNP ID
rs10519911
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:147629439 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCAAAGTCAAGTGATTGTCAATGGA[C/T]CTCTCAAATAAACTGACTTCTAAAT
Phenotype
MIM: 616125 MIM: 613937
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PRMT9 PubMed Links

Gene Details

Gene
PRMT9
Gene Name
protein arginine methyltransferase 9
There are no transcripts associated with this gene.

Gene
TMEM184C
Gene Name
transmembrane protein 184C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018241.2 Intron NP_060711.2

View Full Product Details