Product Details

SNP ID
rs5750604
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:38469606 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTCAGGGGAGGTGGCCAGGAATG[C/T]TGAGTGGGGGACCCGGAGCAGCAGG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KDELR3 PubMed Links

Gene Details

Gene
KDELR3
Gene Name
KDEL endoplasmic reticulum protein retention receptor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006855.3 Intron NP_006846.1
NM_016657.2 Intron NP_057839.1

View Full Product Details