Product Details

SNP ID
rs9397686
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:150146785 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGGGACCCGGATCACAGACTAAAAC[C/T]AAGTGGAGGGGCACTGGTTCTGGAT
Phenotype
MIM: 613242
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PPP1R14C PubMed Links

Gene Details

Gene
PPP1R14C
Gene Name
protein phosphatase 1 regulatory inhibitor subunit 14C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030949.2 Intron NP_112211.1
XM_005267152.4 Intron XP_005267209.1

View Full Product Details