Product Details
- SNP ID
-
rs9932225
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:81244224 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGGTGGAGGCACCAACAGCATTTT[C/G]TGCAGCACCCTGCCTCTAGAGCAGG
- Phenotype
-
MIM: 605748
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
BCO1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs111850415] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- BCO1
- Gene Name
- beta-carotene oxygenase 1
View Full Product Details