Product Details

SNP ID
rs9291317
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.4:47848567 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ATAACAGAAGAGAATATTTCAAACT[G/T]TAGGGCTGACTATGATACTAAATAT
Phenotype
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
LOC101927179 PubMed Links

Gene Details

Gene
LOC101927179
Gene Name
uncharacterized LOC101927179
There are no transcripts associated with this gene.

Gene
NFXL1
Gene Name
nuclear transcription factor, X-box binding like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278623.1 Intron NP_001265552.1
NM_001278624.1 Intron NP_001265553.1
NM_152995.5 Intron NP_694540.3

View Full Product Details