Product Details

SNP ID
rs9633316
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:36799305 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGACAGAAGTGCAAACAGACCCTG[C/G]TGCCCGGCCTTCCTGATTGTCTGTC
Phenotype
MIM: 138243
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GRIK3 PubMed Links

Gene Details

Gene
GRIK3
Gene Name
glutamate ionotropic receptor kainate type subunit 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000831.3 5441 UTR 3 NP_000822.2

View Full Product Details