Product Details

SNP ID
rs10498155
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:223876683 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAATAAAAAAGGTTTGAAAGCTAT[C/G]GAGTAAACTCAGCAGCTGTGATATT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
WDFY1 PubMed Links

Gene Details

Gene
WDFY1
Gene Name
WD repeat and FYVE domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020830.4 3324 UTR 3 NP_065881.1

View Full Product Details