Product Details

SNP ID
rs9658070
Assay Type
DME
NCBI dbSNP Submissions
NA
Location
Chr.6:35347091 on Build GRCh38
Set Membership
DME Validated Inventoried
Context Sequence [VIC/FAM]
GTGTTGTACAGTGTTTTGGGCATGC[A/C]CGTGATACTCACACAGTGGCTTCTG
Phenotype
MIM: 600409
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PPARD PubMed Links

Gene Details

Gene
PPARD
Gene Name
peroxisome proliferator activated receptor delta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171818.1 136 UTR 5 NP_001165289.1
NM_001171819.1 136 UTR 5 NP_001165290.1
NM_001171820.1 136 UTR 5 NP_001165291.1
NM_006238.4 136 UTR 5 NP_006229.1
NM_177435.2 136 UTR 5 NP_803184.1
XM_005249193.1 136 UTR 5 XP_005249250.1
XM_006715120.1 136 UTR 5 XP_006715183.1
XM_006715123.1 136 Intron XP_006715186.1
XM_011514707.1 136 UTR 5 XP_011513009.1
XM_011514709.1 136 UTR 5 XP_011513011.1
XM_011514710.1 136 UTR 5 XP_011513012.1
XM_017010971.1 136 UTR 5 XP_016866460.1
XM_017010972.1 136 UTR 5 XP_016866461.1
XM_017010973.1 136 UTR 5 XP_016866462.1
XM_017010974.1 136 UTR 5 XP_016866463.1
XM_017010975.1 136 Intron XP_016866464.1

View Full Product Details