Product Details

SNP ID
rs5929751
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:136316421 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAAAAACAAACCATTCAGATTGT[C/G]TATCTCTTAGAGTTTTAAACCATTT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ADGRG4 PubMed Links

Gene Details

Gene
ADGRG4
Gene Name
adhesion G protein-coupled receptor G4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153834.3 Intron NP_722576.3

View Full Product Details