Product Details

SNP ID
rs9923279
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:75447513 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTCCATAATGTATTCTTTTGGAG[G/T]ATTCCATAAAGTTTAAGTTCAACAT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
TMEM170A PubMed Links

Gene Details

Gene
TMEM170A
Gene Name
transmembrane protein 170A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304996.1 874 UTR 3 NP_001291925.1
NM_001304997.1 874 UTR 3 NP_001291926.1
NM_001304998.1 874 UTR 3 NP_001291927.1
NM_145254.2 874 UTR 3 NP_660297.1
XM_011522869.2 874 UTR 3 XP_011521171.1
XM_017022939.1 874 UTR 3 XP_016878428.1
XM_017022940.1 874 UTR 3 XP_016878429.1
XM_017022941.1 874 UTR 3 XP_016878430.1
XM_017022942.1 874 Intron XP_016878431.1
XM_017022943.1 874 UTR 3 XP_016878432.1

View Full Product Details