Product Details

SNP ID
rs6074057
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:46174104 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTAGGTGCTTAGTAAGTGAACAGC[C/T]TTCTTTCCTCTTAACTCTGATGGGG
Phenotype
MIM: 609920
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDH22 PubMed Links

Gene Details

Gene
CDH22
Gene Name
cadherin 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021248.2 3491 UTR 3 NP_067071.1
XM_011528994.2 3491 UTR 3 XP_011527296.1
XM_011528995.1 3491 UTR 3 XP_011527297.1

View Full Product Details