Product Details
- SNP ID
-
rs9754334
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:45195711 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATCCCAGGAGTCTAACATCCAACCC[A/G]ACCTATGCAGAATGACGGGTGACCA
- Phenotype
-
MIM: 604646
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
KIAA0930
PubMed Links
Gene Details
- Gene
- KIAA0930
- Gene Name
- KIAA0930
- Gene
- MIR1249
- Gene Name
- microRNA 1249
There are no transcripts associated with this gene.
- Gene
- NUP50
- Gene Name
- nucleoporin 50
There are no transcripts associated with this gene.
View Full Product Details