Product Details
- SNP ID
-
rs9368895
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:35945319 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATTTCTCTGGCTTTTCTAGCTAGAC[A/C]GGATTTCATGACCAACCATTTAAAA
- Phenotype
-
MIM: 608480
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SLC26A8
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs117475331,rs117903426] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SLC26A8
- Gene Name
- solute carrier family 26 member 8
View Full Product Details