Product Details

SNP ID
rs67927428
Assay Type
DME
NCBI dbSNP Submissions
NA
Location
Chr.19:41090365 on Build GRCh38
Set Membership
DME Validated Inventoried
Context Sequence [VIC/FAM]
CTGACTCTCCTCAGACCTCTGAGTT[G/T]ACTCTCTCCCCAACCCCCTTCTCCC
Phenotype
MIM: 608055
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
CYP2A13*1E,g.1894G>T
Literature Links
CYP2A13 PubMed Links

Gene Details

Gene
CYP2A13
Gene Name
cytochrome P450 family 2 subfamily A member 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000766.4 Intron NP_000757.2

View Full Product Details