Product Details

SNP ID
rs11146982
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:132625696 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTCTTCTGTAGAACTTCCACCAG[C/T]GTCATCTCGATGGCAGAGGAGTCGT
Phenotype
MIM: 600844 MIM: 174762
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
P2RX2 PubMed Links

Gene Details

Gene
P2RX2
Gene Name
purinergic receptor P2X 2
There are no transcripts associated with this gene.

Gene
POLE
Gene Name
DNA polymerase epsilon, catalytic subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006231.3 6810 Silent Mutation ACA,ACG T2202T NP_006222.2
XM_011534795.2 6810 Silent Mutation ACA,ACG T2202T XP_011533097.1
XM_011534797.2 6810 Silent Mutation ACA,ACG T1895T XP_011533099.1
XM_011534798.1 6810 Silent Mutation ACA,ACG T1756T XP_011533100.1
XM_011534799.2 6810 Intron XP_011533101.1
XM_011534800.2 6810 Intron XP_011533102.1
XM_011534802.2 6810 Silent Mutation ACA,ACG T1198T XP_011533104.1

View Full Product Details