Product Details
- SNP ID
-
rs12037880
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
7
- Location
-
Chr.1:230324591 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAAGTACAGGTTACCCTCAAAGAAG[G/T]TTCAGTCAATATCTGATTTTGAGTT
- Phenotype
-
MIM: 616791
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
PGBD5
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs117686445] are located under a probe and SNP(s) [rs115109176] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PGBD5
- Gene Name
- piggyBac transposable element derived 5
View Full Product Details