Product Details
- SNP ID
-
rs10902497
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:132108815 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTTTGTTCAAAATACATCTTGAAAC[A/G]TCTTTGTGAAGGCTCTTAGTTTTAA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
EP400NL
PubMed Links
Gene Details
- Gene
- EP400NL
- Gene Name
- EP400 N-terminal like
There are no transcripts associated with this gene.
View Full Product Details