Product Details

SNP ID
rs12348585
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.9:136997917 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCTGGACACTGTGGTTATAGCTCC[G/T]CTTGCTGGTTTCCCACATCCCTGGC
Phenotype
MIM: 600047 MIM: 616315 MIM: 606533
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
ABCA2 PubMed Links

Gene Details

Gene
ABCA2
Gene Name
ATP binding cassette subfamily A member 2
There are no transcripts associated with this gene.

Gene
C9orf142
Gene Name
chromosome 9 open reading frame 142
There are no transcripts associated with this gene.

Gene
CLIC3
Gene Name
chloride intracellular channel 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004669.2 13 Intron NP_004660.2
XM_017015281.1 13 UTR 5 XP_016870770.1
XM_017015282.1 13 Intron XP_016870771.1

View Full Product Details