Product Details

SNP ID
rs11080121
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:30201824 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTGTTCTCAGAAAGGGCTGATATT[C/T]GGCCGGGTGCAGTGGCTCATGCCGG
Phenotype
MIM: 182138
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
SLC6A4 PubMed Links

Gene Details

Gene
SLC6A4
Gene Name
solute carrier family 6 member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001045.5 Intron NP_001036.1

View Full Product Details