Product Details

SNP ID
rs12924052
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:69743343 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACTATAATAACTGTTTCAGGCAAC[C/T]TTAGGTAAGTTCAACGAGAAATAAT
Phenotype
MIM: 613586
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NOB1 PubMed Links

Gene Details

Gene
NOB1
Gene Name
NIN1/PSMD8 binding protein 1 homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014062.2 Intron NP_054781.1

View Full Product Details