Product Details

SNP ID
rs11945310
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:1403509 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCCGCTGCACCTCCCGGTGGGCCC[G/T]GGGCAACGGGCGAGTTCTCGCGGGG
Phenotype
MIM: 610203
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CRIPAK PubMed Links

Gene Details

Gene
CRIPAK
Gene Name
cysteine rich PAK1 inhibitor
There are no transcripts associated with this gene.

Gene
NKX1-1
Gene Name
NK1 homeobox 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290079.1 770 Missense Mutation CAG,CCG Q257P NP_001277008.1

View Full Product Details