Product Details

SNP ID
rs12634316
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:49031896 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAAGGTCAAGGTCAGGAAGCAGG[C/G]CCATGCTCAATATGTGATGAAGGTA
Phenotype
MIM: 146691 MIM: 612911
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
IMPDH2 PubMed Links

Gene Details

Gene
IMPDH2
Gene Name
inosine monophosphate dehydrogenase 2
There are no transcripts associated with this gene.

Gene
NDUFAF3
Gene Name
NADH:ubiquinone oxidoreductase complex assembly factor 3
There are no transcripts associated with this gene.

Gene
QRICH1
Gene Name
glutamine rich 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320580.1 Intron NP_001307509.1
NM_001320581.1 Intron NP_001307510.1
NM_001320582.1 Intron NP_001307511.1
NM_001320583.1 Intron NP_001307512.1
NM_001320584.1 Intron NP_001307513.1
NM_001320585.1 Intron NP_001307514.1
NM_017730.3 Intron NP_060200.2
NM_198880.2 Intron NP_942581.1
XM_011533863.1 Intron XP_011532165.1

View Full Product Details