Product Details

SNP ID
rs12952475
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:47328341 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGTCCTTAAGGACAGAATTTACC[G/T]CTAAACAGAAAGATGGAAACTAAAG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
EFCAB13 PubMed Links

Gene Details

Gene
EFCAB13
Gene Name
EF-hand calcium binding domain 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195192.1 419 UTR 5 NP_001182121.1
NM_152347.4 419 UTR 5 NP_689560.3
Gene
THCAT158
Gene Name
thyroid cancer-associated transcript 158
There are no transcripts associated with this gene.

View Full Product Details