Product Details

SNP ID
rs12301106
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:47664008 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAACTTTACATCACTACTGTAAAT[A/G]AAAAACAGCAATGACACTGATGAAA
Phenotype
MIM: 611477
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RPAP3 PubMed Links

Gene Details

Gene
RPAP3
Gene Name
RNA polymerase II associated protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001146075.1 Intron NP_001139547.1
NM_001146076.1 Intron NP_001139548.1
NM_024604.2 Intron NP_078880.2
XM_011538741.2 Intron XP_011537043.1

View Full Product Details